

2-Aminoazotoluene ≥97%, Sigma-Aldrich®
Catalog # 121568-100G
Leverandør: SIGMA ALDRICH MICROSCOPY
CAS nummer:
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g
- Seq:3401
Specifikationer
Specifikationstestresultater
- Appearance (Color):Orange to Dark Brown
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Wavelength:484 - 490 nm
- Extinction Coefficient:≥2000
- Extinction Coefficient 0.01g/l, 50% Ethanol in 1N HCl:≥17500
- Carbon:≥72.0 %
- Nitrogen:≥17.9 %
- Purity (TLC):≥97 %
- Solubility (Turbidity):Clear
- Solubility (Color), 1mg/ml, Ethanol:Orange to Dark Brown
Specifikationstestresultater
Om denne vare
:
:225.29 g/mol
:M-Clarity™ quality level = MQ200
EINECS:202-591-2
:GHS06
:GHS08
CAS nummer:97-56-3
Formel:C₁₄H₁₅N₃
MDL nummer:MFCD00007733