Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, Benzophenone 100 µg/ml in n-nonane
Organic reference standard, Benzophenone 100 µg/ml in n-nonane
Organic reference standard, Benzophenone 100 µg/ml in n-nonane
CAMBULM-8303-1.2
:
CAMBRIDGE ISOTOPE LABS
:
Organic reference standard, Benzophenone 100 µg/ml in n-nonane
CAMBULM-8303-1.2
:
CAMBRIDGE ISOTOPE LABS
:
Pack type:
Glass bottle
Pk:
1,2 mL
CAS No.:
119-61-9
Description:
Benzophenone 100 µg/ml in n-nonane
Formula:
C₁₃H₁₀O
Description:
Benzophenone