Nickel standard dissolved in oil, 5000 µg/g Ni in mineral oil for wear metal analysis, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Physical Form: Liquid
MDL No.: MFCD00011137
Specifications
- MDL No.:MFCD00011137
Specifications
Related Information
Frequently Bought Together