Forskolin ≥98%
About this item
Purity: 98+% CAS No.: 66575-29-9 Molecular Formula: C22H34O7 Formula Weight: 410.51 Storage Temperature: -30°C to -10°C Physical Form: Solid Solubility: Soluble in DMSO or ethanol. Beilstein Reference: 1692716 Merck Reference: 14,2476 MDL No.: MFCD00082317
An adenylcyclase activator
- CAS Number:66575-29-9
- Formula:C₂₂H₃₄O₇
- MDL Number:MFCD00082317
- Molecular Weight:410.51 g/mol
- Merck Index:14,2476
- EINECS:266-410-9
2 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 66575-29-9 Molecular Formula: C22H34O7 Formula Weight: 410.51 Storage Temperature: -30°C to -10°C Physical Form: Solid Solubility: Soluble in DMSO or ethanol. Beilstein Reference: 1692716 Merck Reference: 14,2476 MDL No.: MFCD00082317
An adenylcyclase activator
- CAS Number:66575-29-9
- Formula:C₂₂H₃₄O₇
- MDL Number:MFCD00082317
- Molecular Weight:410.51 g/mol
- Merck Index:14,2476
- EINECS:266-410-9
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



