Order Entry
Denmark
ContactUsLinkComponent
Retinoic acid, Millipore®
Retinoic acid, Millipore®
Catalog # 554720-500
CAS Number:  
Retinoic acid, Millipore®
Catalog # 554720-500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :300.44 g/mol
EINECS:206-129-0
CAS Number:302-79-4
Formula:C₂₀H₂₈O₂
MDL Number:MFCD00001551