Organic reference standard, Cyanuric acid 100 µg/ml in aqueous solution
Catalog # CAMBULM-8157-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:108-80-5
- Description:Cyanuric acid 100 µg/ml in aqueous solution
- Formula:C₃H₃N₃O₃
- Description:Cyanuric acid