Organic reference standard, 5-Chloro-2-benzoxazolinone 1000 µg/ml in acetonitrile, Cerilliant
Catalog # CERIC-074
Supplier: CERILLIANT
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:95-25-0
- Description:5-Chloro-2-benzoxazolinone 1,000 µg/ml in acetonitrile
- Environmentally Preferable:
- Formula:C₇H₄ClNO₂
- Description:5-Chloro-2-benzoxazolinone