Order Entry
Denmark
ContactUsLinkComponent
Buffers, TRIS, 0,5 M solutions
Buffers, TRIS, 0,5 M solutions
Certifikater
Buffers, TRIS, 0,5 M solutions
Certifikater

Specifikationer

  • CAS No.:
    77-86-1
  • MDL No.:
    MFCD00004679
  • Physical form:
    Liquid

Specifikationer

Frequently Bought Together