Powder funnels, PP, Azlon®
Supplier: SciLabware
Certificates
About this item
Funnels, made of PP, with parallel stem minimises 'bridging' of powder to improve flow.
- Lightweight and unbreakable
- Two largest sizes have vertical sided tops
- Good chemical and temperature resistance
Frequently Bought Together