Tubing connectors, Y-shaped, POM
Supplier: SEMADENI
Certificates
About this item
Connectors, Y-shaped, made of POM, white.
- Temperature resistant up to 100 °C
- Sturdy
- Good chemical resistance
Frequently Bought Together
Connectors, Y-shaped, made of POM, white.