Fernbach culture flasks
Supplier: BELLCO GLASS
Certificates
About this item
These fernbach culture flasks are made of borosilicate glass type 1, class A.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These have three sided baffles with break resistant lip or triple baffled, with 45 mm PP screw cap and pouring ring. For cultures requiring a large surface area to volume ratio.
Frequently Bought Together