Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

High purity dNTPs

Supplier:   Cytiva
High purity dNTPs
Supplier:  Cytiva
Certificates
High purity dNTPs

4 Options Available Below

Product Details & Documents

dNTPs designed for applications requiring high purity such as amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

  • High purity dNTPs are functionally tested in long PCR to be PCR-qualified, and meet or exceed criteria for high-quality sequencing with thermo sequenase DNA polymerase
  • High purity (>98% triphosphate) confirmed by chromatographic analysis and ³¹P nuclear magnetic resonance (NMR) spectroscopy

 : Supplied as solids. For high purity solutions dNTPs, see illustra™ dNTP solutions.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 27-1850-04
  • 27-1860-04
  • 27-1870-04
  • 27-1880-04
  • Purity:
  • ≥ 98% triphosphate
  • ≥ 98% triphosphate
  • ≥ 98% triphosphate
  • ≥ 98% triphosphate
  • Storage conditions:
  • -20°C
  • -20°C
  • -20°C
  • -20°C

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 27-1850-04
    2'-Deoxyadenosine 5'-Triphosphate, disodium, crystalline (dATP)
    2'-Deoxyadenosine 5'-Triphosphate, disodium, crystalline (dATP)
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 3.690,00 kr
    3.690,00 kr
    1 * 250 mg
       3.690,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    27-1850-04
    Purity
    ≥ 98% triphosphate
    Storage conditions
    -20°C
  • 27-1860-04
    2'-Deoxycytidine 5'-Triphosphate, sodium (dCTP)
    2'-Deoxycytidine 5'-Triphosphate, sodium (dCTP)
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 4.340,00 kr
    4.340,00 kr
    1 * 250 mg
       4.340,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    27-1860-04
    Purity
    ≥ 98% triphosphate
    Storage conditions
    -20°C
  • 27-1870-04
    2'-Deoxyguanosine 5'-Triphosphate, sodium (dGTP)
    2'-Deoxyguanosine 5'-Triphosphate, sodium (dGTP)
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 4.340,00 kr
    4.340,00 kr
    1 * 250 mg
       4.340,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    27-1870-04
    Purity
    ≥ 98% triphosphate
    Storage conditions
    -20°C
  • 27-1880-04
    2'-Deoxythymidine 5'-Triphosphate, sodium (dTTP)
    2'-Deoxythymidine 5'-Triphosphate, sodium (dTTP)
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 4.340,00 kr
    4.340,00 kr
    1 * 250 mg
       4.340,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    27-1880-04
    Purity
    ≥ 98% triphosphate
    Storage conditions
    -20°C