Accessories for tangential flow filtration capsules, Minimate™
518-0075
- Description:Minimate™ fitting kit: male Luer to 3,2 mm hose barb, female Luer to 3,2 mm hose barb, 3,2 mm i.d. tubing, tubing screw clamp, tubing clamps, adhesive strips (loop and hook)
- Environmentally Preferable:
- Pk:1