Order Entry
Denmark
ContactUsLinkComponent
Trypsin digestion mix
Trypsin digestion mix
Catalog # 786-242
Supplier:  G-Biosciences
CAS Number:  
Trypsin digestion mix
Catalog # 786-242
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Enzyme name:
    Trypsin digestion mix
  • Grade:
    Mass spectroscopy grade

Specifications

About this item

Trypsin digestion mix provides optimal buffered conditions for in gel trypsin digestion of proteins. Suitable for 100 protein spots.