Ampicillin
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 69-53-4 Molecular Formula: C16H19N3O4S Formula Weight: 349.40 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 208° dec. Solubility: In 1 M NH4OH, 50 mg/ml Beilstein Reference: 1090925 Merck Reference: 14,586 MDL No.: MFCD00005175
- CAS Number:69-53-4
- Formula:C₁₆H₁₉N₃O₄S
- MDL Number:MFCD00005175
- Molecular Weight:349.4 g/mol
- Merck Index:14,586
- Melting Pt:208 °C (dec.)
- EINECS:200-709-7
2 Options Available Below
Product Details & Documents
CAS No.: 69-53-4 Molecular Formula: C16H19N3O4S Formula Weight: 349.40 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 208° dec. Solubility: In 1 M NH4OH, 50 mg/ml Beilstein Reference: 1090925 Merck Reference: 14,586 MDL No.: MFCD00005175
- CAS Number:69-53-4
- Formula:C₁₆H₁₉N₃O₄S
- MDL Number:MFCD00005175
- Molecular Weight:349.4 g/mol
- Merck Index:14,586
- Melting Pt:208 °C (dec.)
- EINECS:200-709-7



