2-Naphthylamine
Glass bottle for solids, 5 g
Catalog # ACTIAS33196G5
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2-Naphthylamine
Catalog # ACTIAS33196G5
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 91-59-8
Jump to:
Product Details & Documents
- CAS Number:91-59-8
- Formula:C₁₀H₉N
- MDL Number:MFCD00004112
- Molecular Weight:143.19 g/mol
- Merck Index:12,06486
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:111…113 °C
- Density:1.0614 g/cm³ (98 °C)
- EINECS:202-080-4
- ADR:6.1,II
- :Density: 1.0614 g/cm³ (98 °C)
- UN:1650
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



