Order Entry
Denmark
Orders LinkContactUsLinkComponent
Galactitol ≥99%

Galactitol ≥99%

Glass bottle for solids, 500 g
Catalog # J63206.36
Galactitol ≥99%
Catalog # J63206.36
CAS Number:  608-66-2
Galactitol ≥99%
Galactitol ≥99%

Glass bottle for solids, 500 g

CAS Number:  608-66-2
Price
3.160,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 608-66-2 Molecular Formula: C6H14O6 Formula Weight: 182.17 Physical Form: Powder Density: 1.47 Appearance: White to off-white Melting Point: 187-191° Boiling Point: 275-280°/1mm Beilstein Reference: 1721903 Merck Reference: 14,4332 MDL No.: MFCD00064288

Useful reagent in bacteriology

Synonyms:  Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:608-66-2
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064288
  • Molecular Weight:182.17 g/mol
  • Merck Index:14,4332
  • Boiling Pt:275…280 °C (1 mmHg)
  • Melting Pt:187…191 °C
  • Density:1.47
  • Storage Temperature:Ambient
  • EINECS:210-165-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications