4,4'-Dimethoxybenzophenone
Glass bottle for solids, 25 g
Catalog # ACTIAS25273G25
Marketsource
Supplier: ACTIVATE SCIENTIFIC
4,4'-Dimethoxybenzophenone
Catalog # ACTIAS25273G25
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 90-96-0
Jump to:
Product Details & Documents
Synonyms: Bis(4-methoxyphenyl)methanone
- CAS Number:90-96-0
- Formula:C₁₅H₁₄O₃
- MDL Number:MFCD00008404
- Molecular Weight:242.27 g/mol
- Boiling Pt:180…240 °C (13 mmHg)
- Melting Pt:142…145 °C
- Density:1.259 g/cm³ (20 °C)
- EINECS:202-028-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



