Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-Bromophenetole

2-Bromophenetole

Glass bottle for solids, 100 g
Catalog # ACTIAS25352G100
Marketsource
Supplier:   ACTIVATE SCIENTIFIC
2-Bromophenetole
Catalog # ACTIAS25352G100
Marketsource
CAS Number:  583-19-7
2-Bromophenetole

Glass bottle for solids, 100 g

CAS Number:  583-19-7
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Bromo-2-ethoxybenzene, 2-Bromophenyl ethyl ether
  • CAS Number:583-19-7
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00061092
  • Molecular Weight:201.06 g/mol
  • EINECS:209-501-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.