Magnesium oxide 96.0-100.5%, powder USP, Macron Fine Chemicals™
6018-23
:
- Pack type:Fiber drum, Leverpack®
- Pk:55 lb
- Seq:7140
- GMP Manufactured Product:—
- Meets U.S.P Requirements:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:—
- USP - Assay (MgO) (ignited basis):96.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acid-Insoluble Substances:≤ 0.1 %
- USP - Arsenic (FCC):≤ 3 mg/kg
- USP - Bulk Density:≤ 0.2 mg/g
- USP - Calcium Oxide (FCC):≤ 1.5 %
- Foreign Matter:≤ 0.2 mg/g
- USP - Lead (Pb)(FCC):≤ 4 mg/kg
- USP - Limit of Calcium (Ca):≤ 1.1 %
- USP - Free Alkali and Soluble Salts:≤ 2.0 %
- USP - Iron (Fe):≤ 0.05 %
- USP - Loss on Ignition:≤ 10.0 %
:
:40.3 g/mol
:13,05700
:3600 °C (1013 hPa)
:2800 °C
:3.58 g/cm³ (25 °C)
:Management of Change (MOC) category = R
:215-171-9
:1309-48-4
:MgO
:MFCD00011109