Order Entry
Denmark
ContactUsLinkComponent
D-(+)-Sucrose ≥99.0% (by HPLC)
D-(+)-Sucrose ≥99.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
D-(+)-Sucrose ≥99.0% (by HPLC)
Supplier:  TCI
Certificates

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Specific rotation [a]20/D:
    +66.3 to +66.7 deg (C=26, H2O)
  • Solubility in hot Water:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together