2-Ethylpyrazine
Glass bottle for solids, 25 g
Catalog # ACTIAS24427G25
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2-Ethylpyrazine
Catalog # ACTIAS24427G25
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 13925-00-3
Jump to:
Product Details & Documents
Synonyms: Ethylpyrazine
- CAS Number:13925-00-3
- Formula:C₆H₈N₂
- MDL Number:MFCD00006149
- Molecular Weight:108.14 g/mol
- Boiling Pt:152 °C (1013 hPa)
- Density:0.98 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:237-691-5
- UN:1993
- ADR:3,III
- :Density: 0.98 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



