Digitonin
Glass bottle, 100 mg
Catalog # TCIAD0540-100MG
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Digitonin
- CAS Number:11024-24-1
- Formula:C₅₆H₉₂O₂₉
- MDL Number:MFCD00077729
- Molecular Weight:1229.312 g/mol
- Merck Index:14,03161
- EINECS:234-255-6
- UN:2811
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystaline
- Specific rotation [a]20/D:-49.0 to -57.0 deg (C=1, EtOH) (after drying)
- Drying loss:max. 8.0 %
- Sensitiveness:to pass test
Specifications
- Pack type:Glass bottle
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



