Order Entry
Denmark
ContactUsLinkComponent
 
D-(+)-Galactosamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
undefined
D-(+)-Galactosamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC) (CAD):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +94.0 to +98.0 deg (C=1, H2O)

 

Frequently Bought Together