


2,4-Dichloro-3-ethyl-6-nitrophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:99817-36-4
:
Formula:C₈H₇Cl₂NO₃
MDL Number:MFCD00270764
ADR:6.1,III
:236.05 g/mol
EINECS:420-740-1
UN:2811
:GHS05
:GHS06
:GHS09
Specification Test Results
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:46.0 to 50.0 deg-C
Specification Test Results
Frequently Bought Together