Pesticide reference standard, 4-Chlorophenol 100 µg/ml in n-nonane
Catalog # CAMBULM-7420-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:106-48-9
- Description:4-Chlorophenol 100 µg/ml in n-nonane
- Formula:C₆H₄ClOH
- Description:4-Chlorophenol