Potassium iodate 1/60 M KIO3 (0.1N) volumetric solution, Fluka™
Catalog # 34273-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:214 g/mol
Melting Pt:560
Density:1.000 g/cm³
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406
Merck Index:13,07726