Order Entry
Denmark
ContactUsLinkComponent
N,N-Dimethylformamide-[carbonyl-¹³C] (99% ¹³C )
Flammable
Health
Irritant
N,N-Dimethylformamide-[carbonyl-¹³C] (99% ¹³C )
Catalog # CAMBCLM-503.1G
CAS Number:  
Flammable
Health
Irritant
N,N-Dimethylformamide-[carbonyl-¹³C] (99% ¹³C )
Catalog # CAMBCLM-503.1G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
UN:2265
ADR:3,III
 :GHS02
 :GHS08
 :GHS07
CAS Number:32488-43-0
Formula:HCOON(CH₃)₂