Tetrabenzyl pyrophosphate ≥98.0% (by HPLC)
TCIAP1223-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:60.0 to 64.0 deg-C
:
:538.48 g/mol
:Freezer
:3261
:8,III
:GHS05
:990-91-0
:C₂₈H₂₈O₇P₂
:MFCD00051941