Order Entry
Denmark
ContactUsLinkComponent
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99.0% (by GC)
Irritant
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥99.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    95.0 to 99.0 deg-C
  • Solubility in hot Methanol:
    within almost transparency

Specification Test Results

Related Information

Frequently Bought Together