Order Entry
Denmark
ContactUsLinkComponent
5-Fluoro-5'-deoxyuridine (Doxifluridine) ≥98.0% (by HPLC, titration analysis)
5-Fluoro-5'-deoxyuridine (Doxifluridine) ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
5-Fluoro-5'-deoxyuridine (Doxifluridine) ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    189.0 to 193.0 deg-C
  • Specific rotation [a]20/D:
    +18.0 to +22.0 deg (C=1, H2O)

Specification Test Results

Related Information

Frequently Bought Together