Order Entry
Denmark
ContactUsLinkComponent
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) ≥99.0% (by HPLC) for biochemical research
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) ≥99.0% (by HPLC) for biochemical research
Supplier:  TCI
Certificates
undefined
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) ≥99.0% (by HPLC) for biochemical research
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Enzyme inhibition test:
    to pass test

Specification Test Results

Related Information

Frequently Bought Together