Order Entry
Denmark
ContactUsLinkComponent
 
2-Dimethylaminoethyl methacrylate ≥98.5% (by GC) stabilised
 :  TCI
 
undefined
2-Dimethylaminoethyl methacrylate ≥98.5% (by GC) stabilised
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.5 %

 

Frequently Bought Together