
2,4-Diaminophenol dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
TCIAD0110-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Gray to Brown powder to crystal
- Purity (with Total Nitrogen):min. 98.0% (calcd.on dried substance)
- Purity (Argentmetric Titration):min. 98.0% (calcd.on dried substance)
- Solubility in Water:very faint turbidity
- Drying loss:max. 3.0 %
:
:197.06 g/mol
:14,02983
:205-279-4
:2811
:6.1,III
:GHS06
:137-09-7
:C₆H₁₀Cl₂N₂O
:MFCD00012979