Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

(±)-Camphorquinone 99%

(±)-Camphorquinone 99%
Certificates
(±)-Camphorquinone 99%

3 Options Available Below

Product Details & Documents
Synonyms:  1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone
  • CAS Number:10373-78-1
  • Formula:C₁₀H₁₄O₂
  • MDL Number:MFCD00064160
  • Molecular Weight:166.22 g/mol
  • Melting Pt:198…202 °C
  • Storage Temperature:Refrigerator
  • EINECS:233-814-1

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    199 to 204 °C
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 554,00 kr
    554,00 kr
    1 * 5 g
       554,00 kr

  • Glass bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 909,00 kr
    909,00 kr
    1 * 10 g
       909,00 kr

  • Glass bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 3.310,00 kr
    3.310,00 kr
    1 * 50 g
       3.310,00 kr