Order Entry
Denmark
ContactUsLinkComponent
2-Bromo-4-formylpyridine ≥95.0% (by GC)
Irritant
2-Bromo-4-formylpyridine ≥95.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
2-Bromo-4-formylpyridine ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to lump
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    53.0 to 57.0 deg-C

Specification Test Results

Frequently Bought Together