3-(Trimethylsilyl)-2-propyn-1-ol ≥95.0% (by GC)
Catalog # TCIAT1500-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
Synonyms: 3-(Trimethylsilyl)propargyl alcohol
:
Boiling Pt:80°C
Flash Pt:67°C
ADR:6.1,III
:128.25 g/mol
EINECS:226-094-5
CAS Number:5272-36-6
Formula:C₆H₁₂OSi
MDL Number:MFCD00002913