Dextran, M.W. ~ 70000 (Dextran 70)
Supplier: TCI
Certificates
About this item
CAS Number:9004-54-0
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935
Merck Index:14,02948
EINECS:232-677-5
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Specific rotation [a]20/D:+170 to +200 deg (C=6, H2O)
- pH:5.0 to 7.0 (60g/L, 25deg-C)
- limiting viscosity:0.21 to 0.26 cp
Specification Test Results
Related Information
Frequently Bought Together