EDTA trisodium salt hydrate ≥98.0% (by titrimetric analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:85715-60-2
:
Formula:C₁₀H₁₃N₂Na₃O₈·H₂O
MDL Number:MFCD00013305
:376.21 g/mol
Storage Temperature:Ambient
EINECS:205-758-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 % (as Dihydrate)
- Water:7.5 to 10.5 %
- Enzyme inhibition test:to pass test
Specification Test Results
Frequently Bought Together