Calcium hydroxide
Catalog # N883-500G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Calcium Carbonate {CaCO3}:15%
- Calcium Hydroxide {Ca(OH)2}:83 %
Specification Test Results
About this item
Synonyms: Slaked lime, Calcium dihydroxide
:
:74.09 g/mol
EINECS:215-137-3
UN:3262
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:1305-62-0
Formula:Ca(OH)₂
MDL Number:MFCD00010901
Melting Pt:550
Density:2.24 g/cm³ (20 °C)
Merck Index:13,01675