

Manganese(II) sulphate monohydrate 98-101%, powder, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:10034-96-5
:
:MnSO₄·1H₂O
:MFCD00149159
:13,05763
:700 °C
:2.95 g/cm³ (25 °C)
:232-089-9
:3077
:9,III
:169.02 g/mol
:GHS08
:GHS09
:Refrigerator
- Meets ACS Reagent Chemical Requirements:98.0 - 101.0%
- For Laboratory,Research,or Manufacturing Use:≤0.01%
- Assay (MnSO₄·H₂O):98.0 - 101.0 %
- Insoluble Matter:≤0.01 %
- Loss on Ignition:10.0 - 12.0 %
- pH of 5% Solution at 25°C:2.0 - 4.0
- Chloride (Cl):≤0.005 %
- Calcium (Ca):≤0.005 %
- Heavy Metals (as Pb):≤0.002 %
- Iron (Fe):≤0.002 %
- Magnesium (Mg):≤0.005 %
- ACS - Nickel (Ni):≤0.02 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.05 %
- Zinc (Zn):≤0.005 %
- Substances Reducing Permanganate:Passes Test
Frequently Bought Together