2-Butyne-1-ol ≥98.0% (by GC)
: TCI
:764-01-2
:
:C₄H₆O
:MFCD00002914
:70.09 g/mol
:145°C
:–2 °C
:0,9483 g/cm³ (20 °C)
:55°C
:Refrigerator
:212-113-4
:1987
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together