Order Entry
Denmark
ContactUsLinkComponent
 
 
Guanidinium hydrochloride ≥99.0% (by titrimetric analysis) for protein research
 :  TCI
 
undefined
 
Guanidinium hydrochloride ≥99.0% (by titrimetric analysis) for protein research
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Melting point:
    185.0 to 189.0 deg-C
  • Absorbance:
    max. 0.200 (6mol/L, H2O, 225.0 nm)
  • Suitability for protein-analysis:
    to pass test

 

Frequently Bought Together