Order Entry
Denmark
ContactUsLinkComponent
 
Urea ≥99.0% (by total nitrogen basis) for biochemical research
 :  TCI
 
undefined
Urea ≥99.0% (by total nitrogen basis) for biochemical research
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 99.0 %
  • Protein denaturation test:
    to pass test
  • Suitability for protein-analysis:
    to pass test

 

Frequently Bought Together