Order Entry
Denmark
ContactUsLinkComponent
Bis(2-ethylhexyl) phthalate-[ring-D₄]
Bis(2-ethylhexyl) phthalate-[ring-D₄]
Catalog # EHERC16173010
Supplier:  EHRENSTORFER
CAS Number:  
Bis(2-ethylhexyl) phthalate-[ring-D₄]
Catalog # EHERC16173010
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:93951-87-2
Formula:C₂₄H₃₄D₄O₄
MDL Number:MFCD00143759