
5-Formylsalicylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:616-76-2
:
Formula:C₈H₆O₄
MDL Number:MFCD00006945
:166.1308 g/mol
EINECS:210-492-0
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together