Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standard, Diethyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  CAMBDLM-1629-1.2
 :  
Organic reference standard, Diethyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  CAMBDLM-1629-1.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    68988-18-1
  • Description:
    Diethyl phthalate-[ring-D4] 100 µg/ml in n-nonane (98% D)
  • Formula:
    C₁₂H₁₄O₄
  • Description:
    Diethyl phthalate-[ring-D4]