Order Entry
Denmark
ContactUsLinkComponent
Di-n-octyl phthalate
Di-n-octyl phthalate
Catalog # EHERC16175000
Supplier:  EHRENSTORFER
CAS Number:  
Di-n-octyl phthalate
Catalog # EHERC16175000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :390.56 g/mol
EINECS:204-214-7
CAS Number:117-84-0
Formula:C₂₄H₃₈O₄
MDL Number:MFCD00015292