Order Entry
Denmark
ContactUsLinkComponent
2-Acrylamido-2-methylpropanesulphonic acid for synthesis, Sigma-Aldrich®
Corrosive
Irritant
2-Acrylamido-2-methylpropanesulphonic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
2-Acrylamido-2-methylpropanesulphonic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Melting Point:
    190 °C
  • Molar Mass:
    207.25 g/mol
  • Flash Point:
    300 °C
  • Density:
    1.36 g/cm3 (20.0 °C)

Specification Test Results

Related Information

Frequently Bought Together