Organic reference standard, Monomethyl phthalate 100 µg/ml in acetonitrile
Catalog # CAMBULM4652-MT-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:4376-18-5
- Description:Monomethyl phthalate 100 µg/ml in acetonitrile
- Formula:C₉H₈O₄
- Description:Monomethyl phthalate